missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Peroxide, 35% w/w, Extra Pure, Scharlab
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Description
- Extra pure
- Synonyms: Hydrogen dioxide, Hydroperoxide
- Molecular Formula: H2O2
- Molar Mass: 34.01g/mol
- CAS: 7722-84-1
- EIN ECS No.: 231-765-0
- Vapor Pressure: ∼20hPa (at 20°C)
- Residue on Evaporation: max 0.05%
Specifications
Specifications
| Boiling Point | ∼110°C |
| Melting Point | ∼ -24°C |
| CAS | 7722-84-1 |
| Physical Form | Solution |
| Quantity | 1 L |
| Molecular Formula | H2O2 |
| Synonym | oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone |
| InChI Key | MHAJPDPJQMAIIY-UHFFFAOYSA-N |
| SMILES | OO |
| IUPAC Name | hydrogen peroxide |
| Show More |
Safety and Handling
Product Identifier
- Hydrogen peroxide, solution 35% w/w (133 vol), extra pure
- Danger
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : Hydrogen Peroxide Solution
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction