Learn More
VEE GEE Hydrometer Jar
The Hydrometer Jar is made of borosilicate glass, the jar features a beaded rim and a circular base. Sized properly to eliminate surface tension effects that lead to inaccurate readings.
Brand:  VEE GEE 21050-600
 View more versions of this product
Product Code. 17816483
  • 20.95 EUR / Each
Estimated Shipment: 07-04-2025
Description
  • The capacity varies depending on the model.
Specifications
Specifications
Flat Circular
15.4 in.
Hydrometer Jar
600 mL
Reusable
20.3 fl. oz.
39 cm
10.8 cm
Borosilicate glass