Learn More
Hydroxylamine Hydrochloride, 96% min., MP Biomedicals
Product Code. p-3547144 Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
Product Code. 11374469
Brand: MP Biomedicals 0210199790
Slowly decomposes when moist
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5470-11-1
Molecular Formula ClH4NO
Molecular Weight (g/mol) 69.49
MDL Number MFCD00051089
InChI Key WTDHULULXKLSOZ-UHFFFAOYSA-N
Synonym hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
PubChem CID 443297
ChEBI CHEBI:5807
IUPAC Name hydroxylamine hydrochloride
SMILES Cl.NO
Specifications
Quantity 500 g
Assay Percent Range 96% min.
Formula Weight 69.5
Chemical Name or Material Hydroxylamine Hydrochloride