Learn More
HOPEN Hygiene PP Coverall with Collar
missing translation for 'orderingAttributeHoverText'
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Color:
Navy
White
Description
- Hygiene PP Coverall
- Classic with collar
- Elasticated
- 40 g/m2
Specifications
Specifications
Product Type | Cleanroom Coveralls |
Size | X-Large |
Material | Polypropylene |
Color | Navy |
Disposable | Single-use |
Sterility | Non-sterile |
Includes | Collar |
For Use With (Application) | Controlled Environments |
Style | Elasticated |