Learn More
Thermo Scientific™ Hypersep™ Hypercarb™ Fixed Well Plate
Unique material for retention of highly polar compounds.
Brand:  Thermo Scientific™ 60306-503
701.20 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 13406986
  • 704.71 EUR / Each
Estimated Shipment: 03-04-2025
Description
Thermo Scientific™ HyperSep™ and Hypercarb™ SPE contains flat,100% porous graphitic carbon (PGC) with selectivity for structurally similar compounds, offering separation of compounds with simple solvents
  • pH stable 0 to 14
  • High batch-to-batch reproducibility
  • Strong retention properties allow use of low bed weights for concentrated extracts
  • Interaction mechanism with polar molecules
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 mg
40 to 60 μm
HypeSep Hypercarb
1/Ea
Universal Vacuum Manifold
HyperSep
96-well Plate