Learn More
Thermo Scientific™ HyperSep™ UniGuard Direct-Connect Online SPE Columns
Product Code. 10187554 Shop All Thermo Scientific Products
missing translation for 'orderingAttributeHoverText'
Sorbent:
HyperSep Hypercarb
HyperSep Quaternary Amine-Modified Polystyrene DVB
HyperSep Sulfonic Acid-Modified Polystyrene DVB
HyperSep Urea-Modified Polystyrene DVB
Diameter (Metric) Inner:
2.1 mm
3.0 mm
missing translation for 'unitSize'
Pack of 4
Product Code. 10187554
missing translation for 'mfr': Thermo Scientific™ 60311202
Only null left
Add to basket
Add to basket
Clean-up and concentrate target analytes in minutes with these direct-connection SPE cartridges, offering quick, easy connection via a reusable drop-in cartridge holder.
Clean-up and concentrate target analytes in minutes with Thermo Scientific™ HyperSep™ Online SPE Products. Available in several convenient formats, the columns utilize popular HyperSep SPE chemistries. When used with conventional HPLC systems, columns and appropriate switching valves, they provide:
  • Effective removal of contaminants, such as proteins, from samples
  • Fast, effective clean-up and concentration of target compounds
Thermo Scientific HyperSep UniGuard Direct-Connect Online SPE Column offer quick, easy connection by means of a reusable drop-in guard cartridge holder.
HyperSep UNIGUARD Direct-Connect Online SPE Cartridges attach directly to the switching valve, just like an HPLC column. They are available with the following HyperSep SPE chemistries:
HyperSep™ Retain™ PEP — for retention of polar and nonpolar analytes
HyperSep Retain-CX — for retention of basic and nonpolar analytes
HyperSep Retain-AX — for retention of acidic and nonpolar analytes
HyperSep™ Hypercarb™ — for retention of extremely polar analytes
TRUSTED_SUSTAINABILITY
Specifications
Description HyperSep Retain PEP; 3.0 mm ID
For Use With (Application) Retention of Polar and Nonpolar Analytes
For Use With (Equipment) Conventional HPLC System
Product Line HyperSep
Sorbent HyperSep Urea-Modified Polystyrene DVB
Type Online Solid Phase Extraction Cartridge
Quantity Pack of 4
Diameter (Metric) Inner 3.0 mm