Learn More
Thermo Scientific™ HyPURITY™ AQUASTAR™ HPLC Columns, 3μm Particle Size
Product Code. 10549365 Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
50 mm
100 mm
150 mm
Diameter (Metric):
2.1 mm
3 mm
4 mm
4.6 mm
Unit Size:
Each
Product Code. 10549365
Brand: Thermo Scientific™ 22503054630
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Polar endcapped for alternative selectivity.
Polar endcapped for alternative selectivity.
  • High quality, highly pure silica columns
Order Info
Other column dimensions are also available. Please call Customer Service for more information.
TRUSTED_SUSTAINABILITY
Specifications
Column Format Analytical Column
Diameter (Metric) 4.6 mm
Length (Metric) 50 mm
Particle Size 3 μm
pH 2 to 8
Pore Size 190 Å
Surface Area 200 m²/g
USP Type L1
Stationary Phase C18
Packing Material Silica
Carbon Load 10%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase