Learn More
Invitrogen™ iBright™ Imaging System Sample Stage/Turntable
Used for sample placement and auto-alignment in the iBright imaging systems
Brand: Invitrogen™ A33829
This item is not returnable.
View return policy
Description
The iBright Imaging System Sample Stage/Turntable is used for sample placement and auto-alignment in the iBright imaging systems (CL750, CL1000, CL1500, FL1000, FL1500). It is made available as a replacement or additional sample stage for these systems. If the iBright imaging system is being used extensively for imaging and band excision from DNA gels, it is recommended that users have a separate Sample Stage to dedicate to this application.Specifications
iBright | |
1 Ea. | |
Imaging System |
Chemiluminescent Western Blot Imaging, DNA Gel Imaging, Protein Gel Imaging | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.