Learn More
BINDER™ ICH Light Module
Shop All BINDER Products
Click to view available options
For Use With (Equipment):
KB PRO 720, KBF/KBF PRO 720
KB PRO/KBF/KBF PRO 260/470
Unit Size:
Each
Description
Module consists of:
- VIS cassette
- UVA cassette
- Independent power supply
Specifications
Specifications
| For Use With (Equipment) | KB PRO 720, KBF/KBF PRO 720 |
| Material | Glass |
| Type | Lighting |