missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGF-I R/IGF1R Mouse anti-Human, Clone: IGF1R/4667, Novus Biologicals™
Mouse Monoclonal Antibody
242.97 EUR - 483.79 EUR
Specifications
Antigen | IGF-I R/IGF1R |
---|---|
Clone | IGF1R/4667 |
Concentration | 0.2 mg/ml |
Dilution | Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin, In vitro assay, Protein Array |
Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), In vitro Assay, Peptide Array |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18331495
|
Novus Biologicals
NBP3-13948-20UG |
20 μg |
242.97 EUR
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18379774
|
Novus Biologicals
NBP3-13948-100UG |
100 μg |
483.79 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IGF-I R/IGF1R Monoclonal antibody specifically detects IGF-I R/IGF1R in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), In vitro Assay, Protein ArraySpecifications
IGF-I R/IGF1R | |
0.2 mg/ml | |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), In vitro Assay, Peptide Array | |
Unconjugated | |
Mouse | |
Apoptosis, Cancer, Lipid and Metabolism, Neuroscience, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
10 mM PBS with 0.05% BSA | |
CD221, CD221 antigen, EC 2.7.10, EC 2.7.10.1, IGF-I receptor, IGFIR, IGFR, insulin-like growth factor 1 receptor, Insulin-like growth factor I receptor, JTK13, MGC142170, MGC142172, MGC18216, soluble IGF1R variant 1, soluble IGF1R variant 2 | |
Recombinant fragment (around aa550-750) of human IGF-I R/IGF1R protein (exact sequence is proprietary) (Uniprot: P08069) | |
Primary | |
Store at 4°C. |
IGF1R/4667 | |
Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin, In vitro assay, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P08069 | |
3480 | |
IgG2c κ | |
Protein A or G purified |