missing translation for 'onlineSavingsMsg'
Learn More

IGHM Mouse anti-Human, Clone: 2D10B10, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66484-1-IG-150UL

 View more versions of this product

Product Code. 16818405

  • 399.00 EUR / 150µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells.
Specifications

Specifications

IGHM
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P01871
IGHM
IGHM Fusion Protein Ag1459
150 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
2D10B10
Unconjugated
IGHM
IGHM, MU, VH
Mouse
Protein G
RUO
3507
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers