missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech IHCeasy Cytokeratin 7 Ready-To-Use IHC Kit
Shop All Proteintech Products
Click to view available options
Quantity:
50 Tests
Unit Size:
1 set
Description
KHC0204 is a ready-to-use IHC kit for staining of Cytokeratin 7(CK7). The kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use. Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Specifications
Specifications
| Clonality | Polyclonal |
| Target Species | Human, Mouse, Rat |
| Quantity | 50 Tests |
| For Use With (Application) | Immunohistochemistry (IHC) |
| Species | Rabbit |
| Content And Storage | All the reagents are stored at 2°C to 8°C. The kit is stable for 6 months from the date of receipt. |
| Kit Contents |
|
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction