Learn More
Proteintech IHCeasy TDP-43 Ready-To-Use IHC Kit
A ready-to-use IHC kit for the staining of TDP43 (also named TARDBP).
Brand:  Proteintech KHC0001-50T
Product Code. 17496703
  • 389.00 EUR / 1 set
Description
  • This kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use.
  • Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Specifications
Specifications
Human, Rat, Mouse
Store at 2°C to 8°C
50 Reactions
FFPE Tissue
50 Tests