Learn More
Proteintech IHCeasy TDP-43 Ready-To-Use IHC Kit
A ready-to-use IHC kit for the staining of TDP43 (also named TARDBP).
Brand: Proteintech KHC0001-50T
This item is not returnable.
View return policy
Description
- This kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use.
- Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Specifications
Human, Rat, Mouse | |
Store at 2°C to 8°C | |
50 Reactions |
FFPE Tissue | |
50 Tests |