Learn More
IKA™ VX 1 Shaker Hand Attachment
Attachment for hand-held vessels
Brand:  IKA 0000607200
Product Code. 10757251
  • 125.00 EUR / Each
In Stock
to see stock.
Explore more special offers
Description
  • For vessels up to 250mL
Specifications
Specifications
One Hand Attachment