Learn More
MMM Medcenter™ Incucell Comfort Line Standard Incubator, 707 L
Temperature Range: 5° above ambient to 99.9°C
Brand: MMM Medcenter™ MC001814
This item is not returnable.
View return policy
Description
- Provided with multiprocessor control system with adaptation for every particular unit type and volume through a Fuzzy-Logic program
- Process information displayed on a clear LCD two-line display
- Control elements placed on a foil keyboard
- 6 adjustable programs
- Chip card system for individual program storage
- RS 232- interface for printer or PC-communication
- Delayed heating start and stop function
- Acoustic and visual alarm in error state
- Time range 0–40 years with 1 min-intervals
- Digital safety thermostat
- Real time
- Programming temperature ramps
- Heating sequences
- Programme cycles
- Manual control of air flap
- Keyboard blocking
- Door opening control
Specifications
707 L | |
Ambient +5° to 99.9°C | |
520 x 940 x 1410 mm |