Learn More
Ismatec™ Independent Channel Control Peristaltic Pump Assembly
Click to view available options
Flow Rate:
35 mL/min.
Unit Size:
Each
Specifications
Specifications
| Channels | 4 |
| Description | 3 stop color coded |
| Enclosure Rating | IP30 |
| Flow Rate | 35 mL/min. |
| Maximum Pressure | 14.5 psi |
| Motor Speed | 0.1 to 100 rpm |
| No. of Rollers | 8 |
| Dimensions (L x W x H) | 182 x 127 x 145 mm |
| Weight (Metric) | 2.7 kg |
| Wattage | 30 w |
| Show More |