missing translation for 'onlineSavingsMsg'
Learn More

Indole-6-carboxylic acid, 98%, Thermo Scientific™

Product Code. 11352857
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1670-82-2
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00210441
InChI Key GHTDODSYDCPOCW-UHFFFAOYSA-N
Synonym indole-6-carboxylic acid, 6-indolecarboxylic acid, 6-carboxyindole, 6-indole carboxylic acid, pubchem1695, 6-carboxy-1h-indole, indole-6-carboxylicacid, indol-6-carboxylic acid, indole 6-carboxylic acid, maybridge4_003509
PubChem CID 595230
IUPAC Name 1H-indole-6-carboxylic acid
SMILES C1=CC(=CC2=C1C=CN2)C(=O)O

Specifications

Melting Point ∽258°C (decomposition)
Quantity 1 g
Beilstein 123991
Formula Weight 161.16
Percent Purity 98%
Chemical Name or Material Indole-6-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'