Learn More
INH14, MedChemExpress
Product Code. 16485626
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16485626
Brand: Medchem Express HY114454
MedChemExpress INH14 is a cell permeable inhibitor of IKKα/IKKβ, with IC50s of 8.97 and 3.59 μM, respectively. INH14 inhibits the IKKα/β-dependent TLR inflammatory response. INH14 also inhibits downstream of TAK1/TAB1 and NF-kB pathways. Anti-inflammatory and anti-cancer activity.
INH14 is a cell permeable inhibitor of IKKα/IKKβ, with IC50s of 8.97 and 3.59 μM, respectively. INH14 inhibits the IKKα/β-dependent TLR inflammatory response. INH14 also inhibits downstream of TAK1/TAB1 and NF-kB pathways. Anti-inflammatory and anti-cancer activity.
Specifications
Chemical Name or Material INH14
Physical Form Solid
CAS 200134-22-1
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C15H16N2O
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : ≥ 125 mg/mL (520.18 mM)
SMILES O=C(NC1=CC=CC=C1)NC2=CC=C(CC)C=C2
Molecular Weight (g/mol) 240.3
Formula Weight 240.3
Percent Purity 98.0%
Grade Research