Learn More
Miele Professional™ Injector Nozzle
Shop All Miele Products
Click to view available options
Model:
ID 110
ID 140
ID 160
ID 180
ID 240
ID 90
Dimensions (Dia. x H):
2.5 dia. x 110mm L
2.5 dia. x 90mm L
4 dia. x 140mm L
4 dia. x 160mm L
4 dia. x 180mm L
6 dia. x 240 mm L
Unit Size:
Each
Description
- Suitable for standard mobile injector units and injector baskets
- Suitable for measuring flasks and test tubes, for example
Specifications
Specifications
| Product Type | Injector Nozzle |
| Model | ID 240 |
| Includes | Plastic support |
| Dimensions (Dia. x H) | 6 dia. x 240 mm L |
| Quantity | 1 Each |