Learn More
WTW™ inoLab™ Multiparameter Digital Benchtop Meters
GREENER_CHOICE
Product Code. 12636637
Click to view available options
Probe Type:
Conductivity Cell
Not included
Optical IDS DO Sensor
pH Electrode, Cond.Cell
pH electrode SenTix 940
pH electrode SenTix 980
Unit Size:
Each
Product Code. 12636637
Brand: WTW™ 1FD354
Only null left
Add to basket
Add to basket
Obtain uncompromised measuring accuracy of digital measurements of pH, ORP, dissolved oxygen (optical), BOD, conductivity and turbidity in the lab using the WTW™ inoLab™ Multiparameter Digital Benchtop Meters.
  • Highly suitable for digital measurements of pH, ORP, dissolved oxygen (optical), BOD, conductivity and turbidity in the lab
  • IDS technology allows optimized measurements and efficient documentation in the simplest manner
  • Offer the correct solution for pH, ORP, dissolved oxygen and conductivity measurements in the lab
  • Uncompromised measuring accuracy
  • Wireless ready, digital sensor recognition, smart sensor evaluation
TRUSTED_SUSTAINABILITY
Specifications
Outputs Mini USB
Accuracy (Conductivity) ±0.05% of reading
Accuracy (DO) ±0.05% of reading
Accuracy (mV) ±0.3 or 0.05% of reading
Accuracy (pH) ±0.005
Calibration Points (Conductivity) 1
Calibration Points (pH) 1 to 5
Certifications/Compliance cETLus, CE
Data Management Details 500 Manual/5000 automatic with time/date stamp
Display Type LCD
Range (Conductivity) 0 μS to 1000 mS
Range (mV) ±1200 mV
Range (pH) -2 to 20.001
Range (Salinity) 0 to 70 ppt
Range (TDS) 0 to 1999 mg/L
Resolution (pH) 0.1/0.01/0.001
Includes Meter with universal power supply, stand and operation manual, optical IDS D.O. sensor FDO™ 925, software and USB cable
Model inoLab Multi 9310 IDS
Power Supply 4 x 1.5V AA type batteries
Probe Type Optical IDS DO Sensor
Product Type Digital benchtop multiparameter
Range (DO) 0 to 90mg/L
Resolution (Conductivity) 3 Significant digits
Resolution (DO) 3 Significant digits
Resolution (mV) 0.1mV
Resolution (Resistivity) 3 Significant digits
Resolution (Salinity) 3 Significant digits
Resolution (TDS) 3 Significant digits
Temperature Accuracy ±0.1
Temperature (Metric) Operating 0°C to 50°C
Temperature Resolution 0.1
Warranty 3 Years
Dimensions (L x W x H) 24 W x 19 Hcm
Width (Metric) 24 cm
Height (Metric) 19 cm
Depth (Metric) 8 cm
Shipping Weight (Metric) 8 kg
Voltage 100/240 V
Frequency 50/60Hz