missing translation for 'onlineSavingsMsg'
Learn More

BD Medical™ Insyte™ Autoguard™ Shielded IV Catheters

Incorporates advanced safety features to help reduce the risk of accidental needlesticks. BD Insyte™ Autoguard™ Shielded IV Catheters are peripheral venous catheters that feature a push button that instantly retracts the needle and a notched needle .

Brand:  BD Medical™ 381812

 View more versions of this product

Product Code. 15594890

  • 81.50 EUR / Pack of 50

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The BD Insyte™ Autoguard™ shielded IV catheter incorporates advanced safety features to reduce accidental sticks, proprietary biomaterials for longer indwelling time, and notched needle technology for first stick success, even in fragile vessels.

  • Safety feature fully encapsulates the entire needle and reduces needlestick injuries by 95%
  • Clinician controlled safety activation technology at the push of a button
  • BD Vialon™ material softens in the vein and exhibits greater kink resistance
  • Increased indwelling time (x 13-16) reduces the total IV costs and supports longer therapies
  • BD Instaflash technology gives immediate confirmation of vessel entry and greater first stick success
  • Quick blood visualization helps to reduce insertion attempts
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BD Vialon
24 G
IV therapies
0.75 in.
Instaflash Notched, Beveled
Yellow
0.7 mm
Autoguard shafety shield
Peripheral Venous Catheter, Shielded
IV Lines
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.