Learn More
Cole-Parmer™ Integrity 10, RS-200, and RS-250 Reaction Station Accessories
missing translation for 'orderingAttributeHoverText'
Description:
Glass Tubes, 24 mm x 150 mm
Integrity Software
Multi IR Box
Multi Temp Module
Non Intrusive IR Sensor
Small Intrusive IR probe PID-NIR5-BNSD
Description
- Suitable for use with Cole-Parmer™ Integrity 10, RS-200, or RS-250 series reaction stations.
- Choose from a wide range of accessories suitable for your reaction station including reaction vessels, probes, and software.
Spécification
Spécification
Description | Glass Tubes, 24 mm x 150 mm |
For Use With (Equipment) | Integrity 10 |
Type | Glass Tubes |