Learn More
Eppendorf™ Interchangeable Platform for Innova™ 5000 Multi-tier Shaker
This interchangeable platform fits shaker model 5000.
Brand: Eppendorf™ M1196-9907
This item is not returnable.
View return policy
Description
- Holds Erlenmeyer flasks or Fernbach flasks
- Dedicated platform
Specifications
6 L Erlenmeyer Flasks | |
Innova™ 5000 Multi-tier Shaker | |
8 |