Learn More
Novus Biologicals™ Intra Acrosomal Protein Overexpression Lysate
Brand: Novus Biologicals™ NBP2-06691
560.00 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
The Intra Acrosomal Protein Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_64454. The protein contains a C-terminal DDK tag. | |
0.1 mg | |
Protein | |
-80°C. Avoid freeze-thaw cycles. |
Western Blot | |
RIPA buffer | |
Homo sapiens Intra Acrosomal Protein, mRNA. | |
Native |
Product Content Correction
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction