Learn More
Invitrogen™ Quant-iT™ RiboGreen™ RNA Reagent
One of most sensitive fluorescent dyes available for quantitation of RNA in solution
Brand:  Invitrogen™ R11491
Product Code. 11503677
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Used with linear fluorescence detection in range of 1-200ng of RNA
  • Available in kit complete with buffer and RNA standard (Cat. No. O11490)
DNA and RNA Purification and Analysis, Nucleic Acid Quantitation, RNA Quantitation
Order Info
Shipping Condition: Room temperature
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only. Not for use in diagnostic procedures.