Learn More
Invitrogen™ Tali™ Calibration Beads
Align and calibrate Tali™ Image-Based Cytometer every three months
Brand: Invitrogen™ T10790
Includes: Tali - Green Calibration Beads, Tali - Red Calibration Beads, Tali - Alignment Beads
Description
Flp-In 293 T-REx cell lines is designed for rapid generation of stable cell lines that ensure homogenous expression of protein of interest from a Flp-In expression vector. Targeted integration of a Flp-In expression vector ensures high-level expression of gene of interest. The Flp-In T-REx™-293 Cell Line contains pFRT/lacZeo and pcDNA™6/TR (from the T-REx™ System) stably integrated. Co-transfection of the Flp-In Cell Lines with a Flp-In expression vector and the Flp recombinase vector, pOG44, results in targeted integration of the expression vector to the same locus in every cell, ensuring homogeneous levels of gene expression.
The Flp-In-293 work well with Flp-In vectors that express a gene from the CMV promoter (e.g., pcDNA™5/FRT, pcDNA5/FRT/V5-His-TOPO™, and pSecTag/FRT/V5-His-TOPO™).
Cell Analysis, Cell Counting and Viability, Cell Counting, Viability, and Cryopreservation, Cell Culture, Cellular Imaging, Imaging Cytometry
Order Info
Shipping Condition: Room temperature
Specifications
Calibration Bead | |
Tali™ - Green Calibration Beads: Store 2 – 6°C Tali™ - Red Calibration Beads: Store 2 – 6°C Tali™ - Alignment Beads; Store 2 – 6°C |
|
Room Temperature | |
Tali™ |
Tali™ Image-Based Cytometer | |
Red, Green | |
Kit | |
1 kit |
For Research Use Only. Not for use in diagnostic procedures.