Learn More
Invitrogen™ UltraPure™ DNase/RNase-Free Distilled Water
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
- UltraPure DNase/RNase-Free Distilled Water is 0.1-μm membrane-filtered and tested for DNase and RNase activity
- Performance and quality testing: No DNase, RNase, or protease activity detected. Our distilled water system is routinely monitored for compliance with current USP monograph test requirements for Water for Injection (WFI)
- Sustainable packaging
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Grade | Molecular Biology |
| Packaging | Bottle |
| Treatment(s) | Not DEPC-Treated |
| Quantity | 500 mL |
| Purification Method | Membrane-Filtered |
| Shipping Condition | Room Temperature |
| Product Line | UltraPure |