missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ Tris Hydrochloride

Simplifies preparation of Tris buffers

Brand:  Invitrogen™ 15506017

146.30 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


Product Code. 11558736

  • 209.00 EUR / 500g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • UltraPure Tris Hydrochloride [tris(hydroxymethyl)aminomethane hydrochloride] used for simplify preparation of Tris buffers

Agarose Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting

Order Info

Shipping Condition: Approved for shipment at room temperature or on wet ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tris
Nucleic Acid Gel Electrophoresis, Blotting
Tris Hydrochloride
Approved for shipment at Room Temperature or on Wet Ice
Store at room temperature.
UltraPure™
500 g
Powder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.