missing translation for 'onlineSavingsMsg'
Learn More

Iodine, Concentrated, For 0.05M/l Solution, Scharlab

Product Code. 11384184
Click to view available options
Quantity:
80 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in analytical chemistry, biochemistry and biology and as a disinfectant.

Concentrated solution of 0.5mol/l ± 0.1% can be used to prepare 1 liter of 0.05mol/l solution containing 12.690g I2.

  • Molecular Formula: I2
  • Molar Mass: 253.81g/mol
  • CAS: 7553-56-2
  • EIN ECS No.: 231-442-4

Specifications

CAS 7553-56-2
Physical Form Solution
Quantity 80 mL
Molecular Formula I2
MDL Number MFCD00011355 MFCD00164163
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
SMILES II
IUPAC Name diiodine
Molecular Weight (g/mol) 253.81
PubChem CID 807
ChEBI CHEBI:17606
Formula Weight 253.81g/mol
Concentration or Composition (by Analyte or Components) 0.5mol/L (concentrated solution)
Density 1.38g/cm3
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'