Learn More
SI Analytics™ Ion-selector Indicator Electrode
Copper electrode with plug head
Brand:  SI Analytics™ 285216273
Product Code. 16346208
  • 1350.00 EUR / Each
Estimated Shipment: 11-03-2025
Description
  • pH range 2 to 6
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Copper Electrode
0.0006 to 6,400 mg/L
2 to 6
0°C to 80°C