Learn More
Memmert™ IQ/OQ Document with Device-Specific Works Test Data
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
WTB11
WTB15
WTB24
WTB35
WTB50
WTB6
Unit Size:
Each
Description
- Includes temperature distribution survey at Memmert
- For 9 measuring points to DIN 12876.2
Specifications
Specifications
| For Use With (Equipment) | WTB35 |
| Product Type | IQ/OQ Document with Device-specific Works Test Data |