Learn More
Memmert™ IQ/OQ Document with Device-Specific Works Test Data
Product Code. 17160385 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
WTB11
WTB15
WTB24
WTB35
WTB50
WTB6
Unit Size:
Each
Product Code. 17160385
Brand: Memmert™ WTB35D00125
Currently Unavailable: We have no information when this product will be available.
View alternative products
For one free-selectable temperature value
  • Includes temperature distribution survey at Memmert
  • For 9 measuring points to DIN 12876.2
Specifications
For Use With (Equipment) WTB35
Product Type IQ/OQ Document with Device-specific Works Test Data