Learn More
Grant Instruments™ IQ/OQ Documentation
Click to view available options
For Use With (Equipment):
All Grant BTD Block Heater Models
All Grant XUB Digital Ultrasonic Baths
All Grant XUBA Analogue Ultrasonic Baths
All QBD Block Heater Models
All QBH Block Heater Models
All SBB Aqua Plus Models
All SUB Aqua Plus Models
CS200G Stirred Thermostatic Bath/Circulator
CXG Stirred Thermostatic Bath/Circulator
Grant CVP-2 PCR Plate Centrifuge
Grant PHMP-100 Thermoshaker for Microplates
Grant R4 Refrigerated Baths/Circulators
Grant T100 Stirred Thermostatic Bath/Circulator
Grant TC120 Stirred Thermostatic Bath/Circulator
Grant TS-DW Deep Well Plate Thermoshaker
Grant TX150 Stirred Thermostatic Bath/Circulator
Grant TXF200 Stirred Thermostatic Bath/Circulator
JBA and SBB Unstirred Baths
JBN/JBA Baths
Unit Size:
Each
Description
- Installation Qualification (IQ): instrument has been delivered and installed in accordance with manufacturer's requirements
- Operational Qualification (OQ): instrument is functioning in accordance with its specification
- Each IQ/OQ documentation set is specific to individual serial numbered equipment and is provided in an easy to use ring bound format.
- It is recommend to purchase IQ/OQ documentation at the time of ordering Grant Scientific equipment.
Specifications
Specifications
| For Use With (Equipment) | All SUB Aqua Plus Models |
| Product Type | IQ/OQ Documentation |