Learn More
Brady™ ISO Safety Sign 'Fire Alarm Flashing Light'
Product Code. 17841765
Click to view available options
Material:
PVC
Shape:
L-shaped
V-shaped
Style:
Class B
Class C
Unit Size:
Each
Product Code. 17841765
Brand: Brady™ 311229
Only null left
Add to basket
Add to basket
ISO Safety Sign displaying flashing fire alarm and flames.
  • To indicate a device that flashes to indicate that the fire alarm has operated
Specifications
Certifications/Compliance Class C
Alternate Part Number F/F018/NT/VPVC-PHOLUMC-151X151-1
Finish Gloss
Material PVC
Quantity 1 Sign
Style Class B
Mounting Type Surface Mount
For Use With (Application) Facility Identification, Safety Identification
Height (English) 5.94 in.
Width (English) 5.94 in.
Temperature Range (Metric) -40°C to +66°C
Shape V-shaped
Temperature Range (English) -40°F to +150°F
Languages Universal
Dimensions (L x W) 151 x 151 mm
Displayed Text Fire Alarm Flashing Light
Product Type Safety Sign
Color Photoluminescent on Red