Learn More
Brady™ ISO Safety Sign without Text - General Warning
Safety sign for hazard identification.
Brand: Brady™ 826611
This item is not returnable.
View return policy
Description
- Sign compliant to ISO 7010
- Improves workplace safety
Specifications
Gloss | |
Permanent Acrylic | |
22 x 25 mm |
Laminated Polyester | |
Triangular | |
Black on Yellow |