Learn More
Brady™ ISO Vertical Safety Sign - Danger: Substances Corrosives
Click to view available options
Dimensions (L x W):
297 x 210 mm
Unit Size:
Each
Description
- Sign compliant to ISO 7010
- Improves workplace safety
Specifications
Specifications
| Alternate Part Number | STFR W023-210x297-PE-CRD/1 |
| Finish | Gloss |
| Material | Laminated Polyester |
| Adhesive Type | Permanent Acrylic |
| Shape | Rectangular |
| Languages | English |
| Dimensions (L x W) | 297 x 210 mm |
| Displayed Text | Danger Substances Corrosives |
| Color | Black on Yellow |