missing translation for 'onlineSavingsMsg'
Learn More

Isobutylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

133.67 EUR - 561.80 EUR

Chemical Identifiers

CAS 926-62-5, 60-29-7
Molecular Formula C4H9BrMg
Molecular Weight (g/mol) 161.33
MDL Number MFCD00075126
InChI Key CMWBEISSZHZIMU-UHFFFAOYSA-M
Synonym isobutylmagnesium bromide, bromo 2-methylpropyl magnesium, magnesium,bromo 2-methylpropyl, isobutylmagnesiumbromide, bromo isobutyl magnesium, iso-butymagnesium bromide, isobutyl magnesium bromide, isobutyl-magnesium bromide, iso-butyl magnesium bromide, 2-methylpropylmagnesiumbromide, grignard reagent
PubChem CID 4285172
IUPAC Name magnesium;2-methanidylpropane;bromide
SMILES CC(C)C[Mg]Br
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10603405
View Documents
Thermo Scientific Acros
427391000
100 mL AcroSeal™ Glass bottle
133.67 EUR
100mL
Please to purchase this item. Need a web account? Register with us today!
10113384
View Documents
Thermo Scientific Acros
427398000
800 mL AcroSeal™ Glass bottle
561.80 EUR
800mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

926-62-5, 60-29-7
161.33
CMWBEISSZHZIMU-UHFFFAOYSA-M
4285172
CC(C)C[Mg]Br
C4H9BrMg
MFCD00075126
isobutylmagnesium bromide, bromo 2-methylpropyl magnesium, magnesium,bromo 2-methylpropyl, isobutylmagnesiumbromide, bromo isobutyl magnesium, iso-butymagnesium bromide, isobutyl magnesium bromide, isobutyl-magnesium bromide, iso-butyl magnesium bromide, 2-methylpropylmagnesiumbromide, grignard reagent
magnesium;2-methanidylpropane;bromide

Specifications

926-62-5, 60-29-7
34.6°C
C4H9BrMg
MFCD00075126
isobutylmagnesium bromide, bromo 2-methylpropyl magnesium, magnesium,bromo 2-methylpropyl, isobutylmagnesiumbromide, bromo isobutyl magnesium, iso-butymagnesium bromide, isobutyl magnesium bromide, isobutyl-magnesium bromide, iso-butyl magnesium bromide, 2-methylpropylmagnesiumbromide, grignard reagent
CC(C)C[Mg]Br
161.33
161.32
0.9410g/mL
−34°C
(CH3)2CHCH2MgBr
0.941
CMWBEISSZHZIMU-UHFFFAOYSA-M
magnesium;2-methanidylpropane;bromide
4285172
Isobutylmagnesium bromide, 2M solution in diethyl ether
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage. May cause drowsiness or dizziness. Harmful if swallowed. Extremely flammable liquid and vapor. In contact with water releases flammable gas. May form explosive peroxides. Reacts violently with water. Repeated exposure may cause skin dryness or cracking.

GHS P Statement
Wear eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Avoid breathing dust/fume/gas/mist/vapors/spray. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Store in a dry place. Store in a closed container.

GHS Signal Word: Danger

Product Identifier
  • Isobutylmagnesium bromide, 2M solution in diethyl ether

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 2

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H261-In contact with water releases flammable gases.
  • H224-Extremely flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.