Learn More
Isobutyltrimethoxysilane, 97%, Thermo Scientific Chemicals
24.40 EUR - 272.00 EUR
Chemical Identifiers
CAS | 18395-30-7 |
---|---|
Molecular Formula | C7H18O3Si |
Molecular Weight (g/mol) | 178.303 |
MDL Number | MFCD00051589 |
InChI Key | XYJRNCYWTVGEEG-UHFFFAOYSA-N |
Synonym | isobutyltrimethoxysilane, isobutyl trimethoxy silane, silane, trimethoxy 2-methylpropyl, trimethoxy 2-methylpropyl silane, dynasylan ibtmo, prosil 178, isobutyltrimethoxy silane, iso-butyl trimethoxysilane, acmc-1cg69, 2-methyl-1-propyltrimethoxysilane |
PubChem CID | 87622 |
IUPAC Name | trimethoxy(2-methylpropyl)silane |
SMILES | CC(C)C[Si](OC)(OC)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11490137
|
Thermo Scientific Alfa Aesar
|
25 g |
24.40 EUR
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11400147
|
Thermo Scientific Alfa Aesar
|
100 g |
66.10 EUR
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11410147
|
Thermo Scientific Alfa Aesar
|
500 g |
272.00 EUR
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18395-30-7 | |
178.303 | |
XYJRNCYWTVGEEG-UHFFFAOYSA-N | |
87622 | |
CC(C)C[Si](OC)(OC)OC |
C7H18O3Si | |
MFCD00051589 | |
isobutyltrimethoxysilane, isobutyl trimethoxy silane, silane, trimethoxy 2-methylpropyl, trimethoxy 2-methylpropyl silane, dynasylan ibtmo, prosil 178, isobutyltrimethoxy silane, iso-butyl trimethoxysilane, acmc-1cg69, 2-methyl-1-propyltrimethoxysilane | |
trimethoxy(2-methylpropyl)silane |
Specifications
154°C to 157°C | |
C7H18O3Si | |
UN1993 | |
43°C (109°F) | |
XYJRNCYWTVGEEG-UHFFFAOYSA-N | |
trimethoxy(2-methylpropyl)silane | |
87622 | |
97% | |
Moisture sensitive | |
Isobutyltrimethoxysilane |
18395-30-7 | |
MFCD00051589 | |
2233976 | |
isobutyltrimethoxysilane, isobutyl trimethoxy silane, silane, trimethoxy 2-methylpropyl, trimethoxy 2-methylpropyl silane, dynasylan ibtmo, prosil 178, isobutyltrimethoxy silane, iso-butyl trimethoxysilane, acmc-1cg69, 2-methyl-1-propyltrimethoxysilane | |
CC(C)C[Si](OC)(OC)OC | |
178.303 | |
178.31 | |
1.396 | |
0.93 |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H336
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 242-272-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only