missing translation for 'onlineSavingsMsg'
Learn More

Isoindoline hydrochloride, 95%, Thermo Scientific™

Product Code. 15409448
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15409448

Brand: Thermo Scientific Alfa Aesar H63012.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Isoindoline hydrochloride is used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Hygroscopic. Store under inder gas. Store away aeay from oxidizing agents, water/ moisture, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32372-82-0
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.63
MDL Number MFCD03502377
InChI Key NOVIRODZMIZUPA-UHFFFAOYSA-N
Synonym isoindoline hydrochloride, 2,3-dihydroisoindole hydrochloride, 2,3-dihydro-1h-isoindole hydrochloride, 2,3-dihydro-1h-isoindole hcl, 1h-isoindole, 2,3-dihydro-, hydrochloride, isoindolinehydrochloride, isoindoline hcl, d0y1rn, dihydroisoindole hydrochloride, ksc491i3f
PubChem CID 12311031
IUPAC Name 2,3-dihydro-1H-isoindole;hydrochloride
SMILES Cl.C1NCC2=CC=CC=C12

Specifications

Melting Point 255°C to 256°C
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 155.62
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Isoindoline hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.