missing translation for 'onlineSavingsMsg'
Learn More

Isoindoline hydrochloride, 95%, Thermo Scientific™

32.20 EUR - 370.00 EUR

Chemical Identifiers

CAS 32372-82-0
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.63
MDL Number MFCD03502377
InChI Key NOVIRODZMIZUPA-UHFFFAOYSA-N
Synonym isoindoline hydrochloride, 2,3-dihydroisoindole hydrochloride, 2,3-dihydro-1h-isoindole hydrochloride, 2,3-dihydro-1h-isoindole hcl, 1h-isoindole, 2,3-dihydro-, hydrochloride, isoindolinehydrochloride, isoindoline hcl, d0y1rn, dihydroisoindole hydrochloride, ksc491i3f
PubChem CID 12311031
IUPAC Name 2,3-dihydro-1H-isoindole;hydrochloride
SMILES Cl.C1NCC2=CC=CC=C12
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15419448
SDS View Documents
Thermo Scientific Alfa Aesar
H63012.MD
250 mg N/A N/A N/A
15499438
SDS View Documents
Thermo Scientific Alfa Aesar
H63012.03
1 g N/A N/A N/A
15409448
SDS View Documents
Thermo Scientific Alfa Aesar
H63012.06
5 g N/A N/A N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Isoindoline hydrochloride is used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Hygroscopic. Store under inder gas. Store away aeay from oxidizing agents, water/ moisture, heat.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

missing translation for 'storageNote' : -30° to -10°C

Recommended Storage : Store at -20°C; Store under Argon

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Isoindoline hydrochloride, 95%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.