missing translation for 'onlineSavingsMsg'
Learn More

Isovaleraldehyde, 98%, Thermo Scientific Chemicals

28.04 EUR - 98.19 EUR

Chemical Identifiers

CAS 590-86-3
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00007014
InChI Key YGHRJJRRZDOVPD-UHFFFAOYSA-N
Synonym isovaleraldehyde, 3-methylbutyraldehyde, isopentaldehyde, isovaleral, isovalerylaldehyde, isoamylaldehyde, butanal, 3-methyl, isopentanal, beta-methylbutanal, isovaleric aldehyde
PubChem CID 11552
ChEBI CHEBI:16638
IUPAC Name 3-methylbutanal
SMILES CC(C)CC=O
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11438860
SDS View Documents
Thermo Scientific Alfa Aesar
A19640.AE
100 mL
28.04 EUR
100mL
Please to purchase this item. Need a web account? Register with us today!
11454187
SDS View Documents
Thermo Scientific Alfa Aesar
A19640.AK
250 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11448860
SDS View Documents
Thermo Scientific Alfa Aesar
A19640.AP
500 mL
65.33 EUR
500mL
Please to purchase this item. Need a web account? Register with us today!
11444187
SDS View Documents
Thermo Scientific Alfa Aesar
A19640.AU
1000 mL
98.19 EUR
1000mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Isovaleraldehyde acts as a reagent in the preparation of active pharmaceutical ingredient (API) products. It serves as an internal standard for the determination of wine aroma carbonyl compounds with 5-8 carbon atoms. Further, it is utilized as a standard to evaluate the quality of olive oils by headspace solid-phase microextraction-gas chromatography using flame ionization detection and multivariate analysis. In addition to this, it is employed to enhance the taste and odor of the food.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isovaleraldehyde acts as a reagent in the preparation of active pharmaceutical ingredient (API) products. It serves as an internal standard for the determination of wine aroma carbonyl compounds with 5-8 carbon atoms. Further, it is utilized as a standard to evaluate the quality of olive oils by headspace solid-phase microextraction-gas chromatography using flame ionization detection and multivariate analysis. In addition to this, it is employed to enhance the taste and odor of the food.

Solubility
Miscible with alcohol and diethyl ether. Slightly miscible with water.

Notes
Light and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, bases, alkaline materials and reducing agents. Store in a cool place.
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Isovaleraldehyde, 98%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.