missing translation for 'onlineSavingsMsg'
Learn More

Itacitinib adipate, MedChemExpress

Product Code. p-200099636
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16487216

Brand: Medchem Express HY16997A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Itacitinib adipate is an orally bioavailable and selective JAK1 inhibitor which has been tested for efficacy and safety in a phase II trial in myelofibrosis.

Itacitinib adipate is an orally bioavailable and selective JAK1 inhibitor which has been tested for efficacy and safety in a phase II trial in myelofibrosis.

Specifications

Chemical Name or Material Itacitinib adipate
CAS 1334302-63-4
Color White
Purity Grade Notes Research
Molecular Formula C32H33F4N9O5
Quantity 100 mg
Solubility Information DMSO : ≥ 137.5 mg/mL (196.52 mM)
SMILES N#CCC1(N2N=CC(C3=C4C(NC=C4)=NC=N3)=C2)CN(C5CCN(C(C6=C(F)C(C(F)(F)F)=NC=C6)=O)CC5)C1.O=C(O)CCCCC(O)=O
Molecular Weight (g/mol) 699.66
Formula Weight 699.66
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.