missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ITD 1
Selective inhibitor of TGF-β signaling; induces cardiomyocyte differentiation in ESCs
295.00 EUR - 1195.00 EUR
Specifications
Purity | >98% |
---|---|
Content And Storage | -20°C |
Product Type | ITD 1 |
Molecular Weight (g/mol) | 415.52 |
Inhibitors | ITD 1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18780499
|
Tocris Bioscience™
5068/50 |
50 mg |
1195.00 EUR
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18769917
|
Tocris Bioscience™
5068/10 |
10 mg |
295.00 EUR
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Selective inhibitor of TGF-β signaling (IC50 = 0.85 μM); displays little or no inhibition of activin, Wnt or BMP signaling pathways. Induces proteasomal degradation of the TGF-β type II receptor. Inhibits TGF-β-induced mesoderm formation from mouse embryonic stem cells (ESCs) during early differentiation; selectively promotes the differentiation of ESCs to cardiomyocytes in vitro between days 3-5. Does not induce differentiation of vascular smooth muscle cells or endothelial cells.Specifications
>98% | |
ITD 1 | |
ITD 1 |
-20°C | |
415.52 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title