Learn More
Avantor™ J.T. Baker™ SAF-T-SPILL™ SOLUSORB™ Solvent Adsorbent Spill Kit
Be prepared for hazardous laboratory spills. For solvent spill clean up, SOLUSORB™ reacts immediately with the solvent, reducing solvent vapors and raising the flash point of the mixture.
Brand: Avantor™ 4437-02
This item is not returnable.
View return policy
Description
- Provides for the safe, effective handling of a solvent spill through reduction of flammable or toxic vapors, fast handling, complete clean up, and convenient disposal
- Each kit is supplied with the appropriate neutraliser, safety glasses, scoops, gloves, disposal bags and ties, and labels.
- Restock your spill control center (or create your own) with replacement kits, available separately.
Specifications
Black | |
Solid | |
1 kit |
Flammable Solvent Spill Kit | |
Solvent Adsorbent, safety glasses, scoops, gloves, disposal bags, ties, and labels |