Learn More
Jujuboside A, MedChemExpress
MedChemExpress Jujuboside A is a glycoside extracted from Semen Ziziphi Spinosae, a Chinese herbal medicine used to treat insomnia and anxiety.
Brand:  Medchem Express HY-N0659
Encompass_Preferred
 View more versions of this product
Product Code. 16439058
  • 113.40 EUR / 10mg
Description
Jujuboside A is a glycoside extracted from Semen Ziziphi Spinosae, a Chinese herbal medicine used to treat insomnia and anxiety.
Specifications
Specifications
Jujuboside A
White
C58H94O26
DMSO : 100 mg/mL (82.83 mM; Need ultrasonic) ∣H2O : 50 mg/mL (41.41 mM; Need ultrasonic)
1207.35
98.0%
Solid
55466-04-1
Research
10 mg
C[C@]([C@]1([H])CC2)(CC[C@]3([H])[C@@]1(CC[C@H](O[C@@](OC[C@H](O)[C@@H]4O[C@@](O[C@H](CO[C@@H]([C@@H]([C@@H](O)[C@@H]5O)O)O[C@@H]5CO)[C@@H](O)[C@@H]6O)([H])[C@@H]6O[C@@](OC[C@@H](O)[C@@H]7O)([H])[C@@H]7O)([H])[C@@H]4O[C@@](O[C@@H](C)[C@H](O)[C@H]8O)([H])[C@@H]8O)C3(C)C)C)[C@@]9(CO%10)[C@@]2([H])[C@]([C@]%11(O)C)([H])[C@@]%10(O[C@@H](/C=C(C)/C)C%11)C9
1207.35
Research
Neuroscience-Neurodegeneration