Learn More
Ansell K200BR Heavy Duty Latex-Coated Industrial Gloves
Product Code. 16875290
Click to view available options
Size:
8
9
10
11
Seamless polycotton gloves with a latex-coating enhance grip capabilities in wet or dry conditions. Open-back design reduces perspiration for comfortable, all day wear.
  • Seamless polycotton gloves are flexible and comfortable
  • Latex-coating enhances grip capabilities in wet or dry conditions
  • Palm & fully thumb coated with a crinkle finish
  • Open back reduces excessive perspiration
TRUSTED_SUSTAINABILITY
Specifications
ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Coating Material Latex
Color Red
Cut Resistance Rating ANSI A2
Exterior Finish Fully Textured
Hand Specific
Material Polycotton
Size 8
EN388 Standard 3241
Glove Cuff Style Knit
Series K200BR