Learn More
WTW™ Kal.Kit Calibration Set for Turb™ 555T Turbidity Meter
Dedicated Calibration Set for the WTW™ Turb™ 555T Turbidity Meter includes indexing rings and cleaning tissues
Brand:  WTW™ 600545
456.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15580665
  • 496.00 EUR / Each
Estimated Shipment: 03-04-2025
Description
  • Calibration set comprising four vials at 0.02, 10.0, 100 and 1750 NTU
  • For use with the Turb™ 555 benchtop advanced laboratory turbidimeter
  • Kit includes tissues and indexing rings
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Turbidity Standard
Turb 555 Turbidity Meter
Four vials at 0.02, 10.0, 100 and 1750 NTU, cleaning tissues and indexing rings