Learn More
Nordvlies Katrin Inclusive Gigant Toilet L Dispenser
Click to view available options
Color:
Black
White
Unit Size:
Each
Description
- Dispenser for big Gigant toilet rolls for high frequency usage
- The lock can be used with or without a key
- The whole paper roll can be used economically with integrated stub roll holder
- Easy paper tear-off to all sides and all edges
- No searching around inside the dispenser opening for the loose end of the paper roll
- Katrin Gigant L Dispensers are suitable for L and M sized Gigant rolls
- New flexible roll brake for better dispensing
- Main raw materials of major plastic parts and lock: ABS (Acrylonitrile butadiene styrene), PC (polycarbonate), POM (polyoxymethylene)
- Katrin Inclusive dispensers are resistant to high temperatures
- Comply with UL94 Fire Safety and Fire Protection regulations (EU)
Specifications
Specifications
| Product Type | Dispenser |
| Color | White |
| Dimensions (L x W x H) | 356 x 342 x 149 mm |
| For Use With (Application) | Dispensing Hand Towels |
| Size | Large |