Learn More
Cole-Parmer™ Keck™ Tubing Adapter
Quick tubing connection without hassle
Brand:  Cole-Parmer™ 06841-50
Product Code. 12736308
  • 66.40 EUR / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Description
  • Safely and easily connect tubing to glassware or other tubing, connect and disconnect with just a quarter twist
  • Unique adapters eliminate broken glass ports and prevent twisted tubing
  • Glassware adapters 06841-51 and -52 are sold separately
Specifications
Specifications
Tubing Adapter
8 to 10 mm
10 Adapters