Learn More
Hamilton™ Custom Kel-F Hub Needles
Design controlled by a series of universal standards which guarantees compatibility between manufacturers.
Brand: Hamilton™ 7750-14
This item is not returnable.
View return policy
Description
- Available in various gauges
- Provided in packs of 6
- Custom point style
- Autoclavable
- Gas sterilizable
- For Kel-F hubs
- Custom length
- Stainless steel Luer tip
Specifications
Autoclavable | |
23 G | |
0.64 mm | |
6/Pk. | |
LT and TLL Syringes and Connectors |
Stainless Steel | |
0.34 mm | |
Luer Lock Kel-F Hub Needle | |
2, 3, or 4 | |
Standard Tapered |