Learn More
Lonza™ KGM™-2 Keratinocyte Growth Medium-2 SingleQuots™ Supplements and Growth Factors
Supplements and growth factors kit for use with keratinocyte basal medium.
Brand: Lonza™ CC-4152
This item is not returnable.
View return policy
Description
- Separately sold SingleQuot™ kit for supplementing basal medium.
- Part of the full KGM™-2 Keratinocyte Growth Medium-2 BulletKit™, which consists of basal medium (CC-3103) and supplements packaged together.
- Includes the following:
- BPE Low Protein - non specific
- HEGF
- Recombinant Human Insulin (0.5%)
- Hydrocortisone Solution
- Transferrin
- Epinephrine
- GA-1000
Specifications
BPE Low Protein - non specific, HEGF, Recombinant Human Insulin 0.5%, Hydrocortisone Solution, Transferrin, Epinephrine, GA-1000 | |
Keratinocyte Cells | |
Supplements and growth factors |